Gene Transcripts

Gene Symbol
Txnip
Entrez Gene ID
117514
Gene Name
thioredoxin interacting protein
Gene Aliases
Vdup1
Chromosome Location
Chr. 2: 198683168 - 198686971 on Build Rnor_6.0
UniGene ID
Rn.2758
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008767.1 NP_001008767.1 8 2252
GenBank mRNA BC088411.1 - 8 2252
U30789.1 - 8 3026

View Full Product Details