Gene Transcripts

Gene Symbol
Kb23
Entrez Gene ID
407759
Gene Name
type II keratin 23
Gene Aliases
Chromosome Location
Chr. 7: 143135074 - 143141634 on Build Rnor_6.0
UniGene ID
Rn.154793
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008813.1 NP_001008813.1 10 1550
XM_006242350.2 XP_006242412.1 9 1486
GenBank mRNA AB355639.1 - 9 1441

View Full Product Details