Gene Transcripts

Gene Symbol
Vom1r8
Entrez Gene ID
494258
Gene Name
vomeronasal 1 receptor 8
Gene Aliases
V1re1
Chromosome Location
Chr. 1: 60117804 - 60118715 on Build Rnor_6.0
UniGene ID
Rn.129457
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008905.1 NP_001008905.1 1 203
GenBank mRNA AY510276.1 - 1 203

View Full Product Details