Gene Transcripts

Gene Symbol
SAMD4B
Entrez Gene ID
55095
Gene Name
sterile alpha motif domain containing 4B
Gene Aliases
SMGB, Smaug2
Chromosome Location
Chr. 19: 39342459 - 39390748 on Build GRCh38
UniGene ID
Hs.612332
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303614.1 NP_001290543.1 7 1749
NM_018028.3 NP_060498.2 10 2470
XM_011527057.2 XP_011525359.1 8 1760
XM_011527060.2 XP_011525362.1 8 2862
XM_011527061.2 XP_011525363.1 7 1674
XM_011527062.2 XP_011525364.1 7 1735
XM_011527063.2 XP_011525365.1 8 1759
XM_011527064.2 XP_011525366.1 8 1760
XM_017026918.1 XP_016882407.1 7 1694
XM_017026919.1 XP_016882408.1 9 1850
XM_017026920.1 XP_016882409.1 9 1872
XM_017026921.1 XP_016882410.1 7 1735
XM_017026922.1 XP_016882411.1 9 1852
XM_017026923.1 XP_016882412.1 9 7390
GenBank mRNA BC039652.1 - 3 447
BC054518.1 - 10 2461 6501489
BC065211.1 - 4 757
BC080641.1 - 7 1709 6645022
EF601121.1 - 10 2457

View Full Product Details