Gene Transcripts

Gene Symbol
Mrgpre
Entrez Gene ID
404660
Gene Name
MAS-related GPR, member E
Gene Aliases
Mgrf, MrgE
Chromosome Location
Chr. 1: 216967899 - 216973906 on Build Rnor_6.0
UniGene ID
Rn.189891
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002288.1 NP_001002288.1 1 257
XM_006230870.2 XP_006230932.1 4 823
XM_006230872.2 XP_006230934.1 2 621
XM_008760163.1 XP_008758385.1 3 719

View Full Product Details