Gene Transcripts

Gene Symbol
Nenf
Entrez Gene ID
289380
Gene Name
neudesin neurotrophic factor
Gene Aliases
SCIRP10, Spuf
Chromosome Location
Chr. 13: 109990294 - 109997092 on Build Rnor_6.0
UniGene ID
Rn.3355
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002851.1 NP_001002851.1 4 558
GenBank mRNA AY623793.1 - 4 558

View Full Product Details