Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001004062.2 | NP_001004062.1 | 14 | 2468 | |
GenBank mRNA | AK129173.1 | - | 14 | 2465 | |
BC056137.1 | - | 14 | 7056 | ||
BC068139.1 | - | 14 | 2474 | ||
BC080308.1 | - | 14 | 2468 | 5719562 |