Gene Transcripts

Gene Symbol
OR13G1
Entrez Gene ID
441933
Gene Name
olfactory receptor family 13 subfamily G member 1
Gene Aliases
OR1-37
Chromosome Location
Chr. 1: 247672118 - 247673041 on Build GRCh38
UniGene ID
Hs.553834
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005487.1 NP_001005487.1 1 52
GenBank mRNA BC136736.1 - 1 74
BC136738.1 - 1 74 9020728

View Full Product Details