Gene Transcripts

Gene Symbol
Qtrtd1
Entrez Gene ID
106248
Gene Name
queuine tRNA-ribosyltransferase domain containing 1
Gene Aliases
3110012M05Rik, 4930470H18Rik, AI648807
Chromosome Location
Chr. 16: 43861407 - 43889680 on Build GRCm38
UniGene ID
Mm.275354
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029128.2 NP_083404.2 2 145
GenBank mRNA AK014045.1 - 2 126
AK015536.1 - 158
BC017628.1 - 2 117
BC083089.1 - 2 125 30105859
BY020492.1 - 2 145
FM985972.1 - 1 25

View Full Product Details