Gene Transcripts

Gene Symbol
POTEA
Entrez Gene ID
340441
Gene Name
POTE ankyrin domain family member A
Gene Aliases
A26A1, CT104.3, POTE-8, POTE8
Chromosome Location
Chr. 8: 43292370 - 43363185 on Build GRCh38
UniGene ID
Hs.531579
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002920.1 NP_001002920.1 10 1161
NM_001005365.2 NP_001005365.2 11 1299
GenBank mRNA AY462869.1 - 11 1299
AY462870.1 - 10 1161

View Full Product Details