Gene Transcripts

Gene Symbol
Ndufa11
Entrez Gene ID
301123
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 11
Gene Aliases
Chromosome Location
Chr. 9: 10334104 - 10338163 on Build Rnor_6.0
UniGene ID
Rn.96380
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212517.1 NP_997682.1 2 163
GenBank mRNA AY272059.1 - 2 163
FQ209810.1 - 2 143

View Full Product Details