Gene Transcripts
- Gene Symbol
- FAM19A1
- Entrez Gene ID
- 407738
- Gene Name
- family with sequence similarity 19 member A1, C-C motif chemokine like
- Gene Aliases
- TAFA-1, TAFA1
- Chromosome Location
- Chr. 3: 67991592 - 68546056 on Build GRCh38
- UniGene ID
- Hs.655061
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252216.1 |
NP_001239145.1
|
4, 5 |
604 |
|
NM_213609.3 |
NP_998774.2
|
4, 5 |
878 |
|
XM_017006386.1 |
XP_016861875.1
|
3, 4 |
1025 |
|
XM_017006387.1 |
XP_016861876.1
|
4, 5 |
1517 |
|
XM_017006388.1 |
XP_016861877.1
|
4, 5 |
1310 |
|
XM_017006389.1 |
XP_016861878.1
|
3, 4 |
1022 |
|
XM_017006390.1 |
XP_016861879.1
|
4, 5 |
1518 |
|
GenBank mRNA |
AK289668.1 |
- |
4, 5 |
867 |
|
AK290446.1 |
- |
4, 5 |
800 |
|
AL713702.1 |
- |
3, 4 |
787 |
|
AY325114.1 |
- |
4, 5 |
604 |
|
BC025746.1 |
- |
4, 5 |
780 |
5200142 |
View Full Product Details