Gene Transcripts

Gene Symbol
DMXL2
Entrez Gene ID
23312
Gene Name
Dmx like 2
Gene Aliases
PEPNS, RC3
Chromosome Location
Chr. 15: 51447724 - 51622833 on Build GRCh38
UniGene ID
Hs.511386
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174116.1 NP_001167587.1 2 333
NM_001174117.1 NP_001167588.1 2 333
NM_015263.3 NP_056078.2 2 333
XM_005254255.1 XP_005254312.1 2 396
XM_005254256.1 XP_005254313.1 2 396
XM_011521402.2 XP_011519704.1 2 396
XM_017022034.1 XP_016877523.1 2 69
GenBank mRNA AF389880.1 - 2 315
AK000156.1 - 2 334
BC140781.1 - 2 198 9021649
BC144539.1 - 2 198 9053066
BX491117.1 - 2 214
DA484900.1 - 2 396

View Full Product Details