Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001323614.1 | NP_001310543.1 | 5 | 704 | |
NM_001323615.1 | NP_001310544.1 | 5 | 704 | ||
NM_001323616.1 | NP_001310545.1 | 6 | 836 | ||
NM_001323617.1 | NP_001310546.1 | 6 | 671 | ||
NM_015327.2 | NP_056142.2 | 5 | 643 | ||
XM_017000843.1 | XP_016856332.1 | 5 | 650 | ||
GenBank mRNA | AB029012.2 | - | 5 | 645 | |
AB085691.1 | - | 5 | 571 | ||
AK092183.1 | - | 5 | 619 | ||
AY168922.1 | - | 5 | 497 | ||
AY336728.1 | - | 5 | 494 | ||
BC038296.1 | - | 5 | 644 | 4824487 | |
CT000143.1 | - | 5 | 704 |