Gene Transcripts
- Gene Symbol
- SCAP
- Entrez Gene ID
- 22937
- Gene Name
- SREBF chaperone
- Gene Aliases
- Chromosome Location
- Chr. 3: 47413685 - 47477126 on Build GRCh38
- UniGene ID
- Hs.531789
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320044.1 |
NP_001306973.1
|
7, 8 |
1027 |
|
NM_012235.3 |
NP_036367.2
|
10, 11 |
1512 |
|
XM_005264967.1 |
XP_005265024.1
|
10, 11 |
1439 |
|
XM_005264968.1 |
XP_005265025.1
|
7, 8 |
1181 |
|
XM_005264971.1 |
XP_005265028.1
|
4, 5 |
592 |
|
XM_011533501.1 |
XP_011531803.1
|
10, 11 |
1539 |
|
XM_011533502.2 |
XP_011531804.1
|
2, 3 |
188 |
|
XM_017005918.1 |
XP_016861407.1
|
10, 11 |
1526 |
|
XM_017005919.1 |
XP_016861408.1
|
10, 11 |
1459 |
|
XM_017005920.1 |
XP_016861409.1
|
10, 11 |
1459 |
|
XM_017005921.1 |
XP_016861410.1
|
4, 5 |
592 |
|
GenBank mRNA |
AK027402.1 |
- |
4, 5 |
505 |
|
AK075528.1 |
- |
2, 3 |
206 |
|
AK291484.1 |
- |
5, 6 |
705 |
|
AK300311.1 |
- |
7, 8 |
917 |
|
BC020987.1 |
- |
2, 3 |
233 |
3852258 |
BC027207.2 |
- |
6, 7 |
711 |
|
D83782.2 |
- |
10, 11 |
1498 |
|
View Full Product Details