Gene Transcripts
- Gene Symbol
- PIGT
- Entrez Gene ID
- 51604
- Gene Name
- phosphatidylinositol glycan anchor biosynthesis class T
- Gene Aliases
- CGI-06, MCAHS3, NDAP, PNH2
- Chromosome Location
- Chr. 20: 45416067 - 45426245 on Build GRCh38
- UniGene ID
- Hs.437388
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001184728.2 |
NP_001171657.1
|
8, 9 |
1151 |
|
NM_001184730.2 |
NP_001171659.1
|
7, 8 |
1013 |
|
NM_015937.5 |
NP_057021.2
|
9, 10 |
1319 |
|
NR_047691.1 |
|
9, 10 |
1279 |
|
NR_047692.1 |
|
8, 9 |
1222 |
|
NR_047693.1 |
|
8, 9 |
1218 |
|
NR_047694.1 |
|
8, 9 |
1141 |
|
NR_047695.1 |
|
6, 7 |
912 |
|
XM_005260432.2 |
XP_005260489.1
|
8, 9 |
1113 |
|
GenBank mRNA |
AB057724.1 |
- |
9, 10 |
1229 |
|
AF132940.1 |
- |
9, 10 |
1223 |
|
AK075469.1 |
- |
9, 10 |
1243 |
|
AK293294.1 |
- |
8, 9 |
1142 |
|
AK293686.1 |
- |
8, 9 |
1064 |
|
AK296063.1 |
- |
8, 9 |
1148 |
|
AK296139.1 |
- |
8, 9 |
1074 |
|
AK297830.1 |
- |
4, 5 |
798 |
|
AK299013.1 |
- |
6, 7 |
838 |
|
AK302093.1 |
- |
7, 8 |
971 |
|
AK311100.1 |
- |
8, 9 |
1101 |
|
AK316358.1 |
- |
9, 10 |
1205 |
|
AL121742.1 |
- |
9, 10 |
1222 |
|
AY358588.1 |
- |
9, 10 |
1226 |
|
BC015022.1 |
- |
9, 10 |
1248 |
3921680 |
BC110892.1 |
- |
9, 10 |
1227 |
|
BC136827.1 |
- |
9, 10 |
1257 |
9020818 |
BC136828.1 |
- |
9, 10 |
1257 |
9020819 |
View Full Product Details