Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016089.2 | NP_057173.2 | 3 | 188 | |
XM_017006575.1 | XP_016862064.1 | 3 | 272 | ||
XM_017006576.1 | XP_016862065.1 | 5 | 418 | ||
XM_017006577.1 | XP_016862066.1 | 3 | 1474 | ||
XM_017006578.1 | XP_016862067.1 | 3 | 252 | ||
XM_017006579.1 | XP_016862068.1 | 5 | 377 | ||
GenBank mRNA | AB209387.1 | - | 3 | 169 | |
AF114816.1 | - | 5 | 494 | ||
AF114817.1 | - | 5 | 494 | ||
AK298782.1 | - | 3 | 188 | ||
AL832831.1 | - | 3 | 173 | ||
AY258146.1 | - | 3 | 188 | ||
BC005859.2 | - | 3 | 147 | ||
BC006247.2 | - | 3 | 168 | ||
BC048798.1 | - | 3 | 165 |