Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317851.1 | NP_001304780.1 | 7 | 931 | |
NM_016173.4 | NP_057257.1 | 7 | 1232 | ||
XM_011533806.2 | XP_011532108.1 | 7 | 1232 | ||
XM_011533807.1 | XP_011532109.1 | 7 | 1117 | ||
XM_011533808.1 | XP_011532110.1 | 7 | 903 | ||
XM_011533809.2 | XP_011532111.1 | 7 | 1232 | ||
XM_011533810.2 | XP_011532112.1 | 7 | 1232 | ||
XM_011533813.2 | XP_011532115.1 | 7 | 1270 | ||
XM_011533814.2 | XP_011532116.1 | 7 | 1194 | ||
GenBank mRNA | AF131220.1 | - | 6 | 818 | |
AF172244.1 | - | 7 | 832 | ||
AK025973.1 | - | 7 | 931 | ||
AK314023.1 | - | 7 | 852 | ||
BC000781.2 | - | 6 | 800 | 3505225 | |
BX537562.1 | - | 7 | 868 |