Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145779.1 | NP_001139251.1 | 7, 8 | 690 | |
NM_008442.2 | NP_032468.2 | 8, 9 | 1073 | ||
GenBank mRNA | AK016720.1 | - | 7, 8 | 693 | |
AK037037.1 | - | 7, 8 | 603 | ||
BC006803.1 | - | 7, 8 | 685 | 3152883 | |
D12644.1 | - | 8, 9 | 1074 | ||
Y15894.1 | - | 6, 7 | 540 |