Gene Transcripts
- Gene Symbol
- Mapk14
- Entrez Gene ID
- 26416
- Gene Name
- mitogen-activated protein kinase 14
- Gene Aliases
- CSBP2, Crk1, Csbp1, Mxi2, PRKM14, PRKM15, p38, p38-alpha, p38MAPK, p38a, p38alpha
- Chromosome Location
- Chr. 17: 28691342 - 28748405 on Build GRCm38
- UniGene ID
- Mm.311337
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001168513.1 |
NP_001161985.1
|
8, 9 |
688 |
|
NM_001168514.1 |
NP_001161986.1
|
9, 10 |
1132 |
|
NM_011951.3 |
NP_036081.1
|
8, 9 |
1087 |
|
GenBank mRNA |
AF128892.1 |
- |
8, 9 |
985 |
|
AK133684.1 |
- |
8, 9 |
1000 |
|
AK151348.1 |
- |
8, 9 |
689 |
|
AK153025.1 |
- |
8, 9 |
689 |
|
AK178357.1 |
- |
|
28 |
|
BC012235.1 |
- |
8, 9 |
1087 |
4195842 |
BC018227.1 |
- |
9, 10 |
1029 |
|
U10871.1 |
- |
8, 9 |
985 |
|
View Full Product Details