Gene Transcripts

Gene Symbol
OR13D1
Entrez Gene ID
286365
Gene Name
olfactory receptor family 13 subfamily D member 1
Gene Aliases
OR9-15
Chromosome Location
Chr. 9: 104694422 - 104695462 on Build GRCh38
UniGene ID
Hs.553683
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004484.1 NP_001004484.1 1 904
GenBank mRNA BC140916.1 - 1 947

View Full Product Details