Gene Transcripts

Gene Symbol
ANAPC7
Entrez Gene ID
51434
Gene Name
anaphase promoting complex subunit 7
Gene Aliases
APC7
Chromosome Location
Chr. 12: 110372900 - 110403730 on Build GRCh38
UniGene ID
Hs.719935
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137664.1 NP_001131136.1 5 663
NM_016238.2 NP_057322.2 5 663
XM_005253903.3 XP_005253960.1 4 529
XM_011538454.2 XP_011536756.1 5 891
XM_017019425.1 XP_016874914.1 5 663
XM_017019426.1 XP_016874915.1 5 658
XM_017019427.1 XP_016874916.1 5 1457
XM_017019428.1 XP_016874917.1 4 546
GenBank mRNA AF191340.1 - 5 560
AK021809.1 - 5 642
BC009498.2 - 3 233
BC040130.1 - 5 578
BC098264.1 - 5 559
BC098295.1 - 5 559
BC098362.1 - 5 559
BC099732.1 - 5 559
BC111798.1 - 5 559
BC141848.1 - 5 613 8860346
BC148236.2 - 5 560 40024981

View Full Product Details