Gene Transcripts

Gene Symbol
SUDS3
Entrez Gene ID
64426
Gene Name
SDS3 homolog, SIN3A corepressor complex component
Gene Aliases
SAP45, SDS3
Chromosome Location
Chr. 12: 118373189 - 118418035 on Build GRCh38
UniGene ID
Hs.416630
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022491.2 NP_071936.2 8 784
XM_011538671.2 XP_011536973.1 9 1861
XM_017019817.1 XP_016875306.1 7 668
XM_017019818.1 XP_016875307.1 10 1016
XM_017019819.1 XP_016875308.1 9 878
XM_017019820.1 XP_016875309.1 10 910
GenBank mRNA AK023801.1 - 3 260
AK024460.1 - 8 767
BC030252.1 - 2 157
BC093990.1 - 3 214 7939835
BC098404.1 - 8 807 30332266
BC112000.1 - 3 214
BX647687.1 - 9 742

View Full Product Details