Gene Transcripts

Gene Symbol
LMF1
Entrez Gene ID
64788
Gene Name
lipase maturation factor 1
Gene Aliases
C16orf26, HMFN1876, JFP11, TMEM112, TMEM112A
Chromosome Location
Chr. 16: 853632 - 981615 on Build GRCh38
UniGene ID
Hs.71912
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022773.2 NP_073610.2 9 1323
NR_033645.1 10 1452
NR_033646.1 11 1630
XM_006720928.2 XP_006720991.1 9 1596
XM_006720929.3 XP_006720992.1 10 1477
XM_011522614.2 XP_011520916.1 9 1321
XM_011522617.2 XP_011520919.1 7 972
XM_017023575.1 XP_016879064.1 9 1321
XM_017023576.1 XP_016879065.1 8 1087
XM_017023577.1 XP_016879066.1 10 1752
XM_017023578.1 XP_016879067.1 8 1172
XM_017023579.1 XP_016879068.1 8 1180
XM_017023580.1 XP_016879069.1 11 1902
GenBank mRNA AB075873.1 - 9 1307
AK022743.1 - 9 1321
AK025955.1 - 9 1255
AK093050.1 - 6 830
AK125201.1 - 10 1454
AK225065.1 - 9 1321
AK225406.1 - 9 1308
AL832177.1 - 3 2941
BC010738.1 - 8 1088

View Full Product Details