Gene Transcripts
- Gene Symbol
- MFSD14B
- Entrez Gene ID
- 84641
- Gene Name
- major facilitator superfamily domain containing 14B
- Gene Aliases
- HIATL1
- Chromosome Location
- Chr. 9: 94374327 - 94460920 on Build GRCh38
- UniGene ID
- Hs.555996
- Species
- Homo sapiens
- Gene Symbol
- MFSD14C
- Entrez Gene ID
- 84278
- Gene Name
- major facilitator superfamily domain containing 14C
- Gene Aliases
- HIATL2
- Chromosome Location
- Chr. 9: 96946045 - 97013580 on Build GRCh38
- UniGene ID
- Hs.610084
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002894.1 |
|
4 |
592 |
|
GenBank mRNA |
BC005058.1 |
- |
4 |
592 |
|
View Full Product Details