Gene Transcripts

Gene Symbol
TRAT1
Entrez Gene ID
50852
Gene Name
T cell receptor associated transmembrane adaptor 1
Gene Aliases
HSPC062, TCRIM, TRIM, pp29/30
Chromosome Location
Chr. 3: 108822698 - 108855005 on Build GRCh38
UniGene ID
Hs.138701
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317747.1 NP_001304676.1 4, 5 430
NM_016388.3 NP_057472.2 5, 6 541
GenBank mRNA AF161547.1 - 5, 6 456
AJ224878.1 - 5, 6 440
AJ240085.1 - 4, 5 356
AK291991.1 - 5, 6 541
BC025713.1 - 5, 6 447 5227396
BX415345.2 - 5, 6 449

View Full Product Details