Gene Transcripts
- Gene Symbol
- OTUD4
- Entrez Gene ID
- 54726
- Gene Name
- OTU deubiquitinase 4
- Gene Aliases
- DUBA6, HIN1, HSHIN1
- Chromosome Location
- Chr. 4: 145133650 - 145180019 on Build GRCh38
- UniGene ID
- Hs.270851
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001102653.1 |
NP_001096123.1
|
5, 6 |
365 |
|
NM_017493.6 |
NP_059963.1
|
4, 5 |
263 |
|
XM_005263079.4 |
XP_005263136.1
|
5, 6 |
468 |
|
XM_011532040.2 |
XP_011530342.2
|
5, 6 |
469 |
|
XM_011532041.2 |
XP_011530343.2
|
5, 6 |
469 |
|
XM_011532042.2 |
XP_011530344.1
|
5, 6 |
390 |
|
GenBank mRNA |
AK302581.1 |
- |
5, 6 |
327 |
|
BC118572.1 |
- |
4, 5 |
259 |
40036249 |
BC118653.1 |
- |
4, 5 |
259 |
40036248 |
BM471766.1 |
- |
2, 3 |
118 |
|
DB446926.2 |
- |
5, 6 |
367 |
|
X68242.1 |
- |
5, 6 |
354 |
|
View Full Product Details