Gene Transcripts

Gene Symbol
Hps5
Entrez Gene ID
246694
Gene Name
Hermansky-Pudlak syndrome 5 homolog (human)
Gene Aliases
AI646796, AL022647, C85120, haze, hz, maroon, mr, ru-2, ru2
Chromosome Location
Chr. 7: 46760466 - 46796064 on Build GRCm38
UniGene ID
Mm.379442
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005247.2 NP_001005247.2 5 936
NM_001005248.2 NP_001005248.2 5 912
NM_001167864.1 NP_001161336.1 5 936
GenBank mRNA AF534396.1 - 5 733
AF534397.1 - 5 709
AF534398.1 - 3 298
AF534399.1 - 5 733
AK129265.1 - 5 863
AK141088.1 - 5 734
AK154612.1 - 5 754
BC046443.1 - 5 551
BC082542.1 - 5 936 6405811

View Full Product Details