Gene Transcripts

Gene Symbol
Olr13
Entrez Gene ID
293075
Gene Name
olfactory receptor 13
Gene Aliases
Chromosome Location
Chr. 1: 149786382 - 149787308 on Build Rnor_6.0
UniGene ID
Rn.141953
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000116.1 NP_001000116.1 1 271

Gene Symbol
LOC100909966
Entrez Gene ID
100909966
Gene Name
olfactory receptor 6F1-like
Gene Aliases
Chromosome Location
Chr. 1: 149602346 - 149603284 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748916.2 XP_003748964.1 283

View Full Product Details