Gene Transcripts

Gene Symbol
Lrrc2
Entrez Gene ID
301033
Gene Name
leucine rich repeat containing 2
Gene Aliases
Chromosome Location
Chr. 8: 119228533 - 119259649 on Build Rnor_6.0
UniGene ID
Rn.74973
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012001.1 NP_001012001.1 2499
GenBank mRNA BC085884.1 - 2499

View Full Product Details