Gene Transcripts

Gene Symbol
CWC25
Entrez Gene ID
54883
Gene Name
CWC25 spliceosome associated protein homolog
Gene Aliases
CCDC49
Chromosome Location
Chr. 17: 38800434 - 38825350 on Build GRCh38
UniGene ID
Hs.406223
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017748.4 NP_060218.1 2 268
NR_073428.1 2 268
GenBank mRNA AK000298.1 - 2 218
AK225373.1 - 2 239
AK296115.1 - 2 233
AK302983.1 - 2 169
BC003085.1 - 2 121
BC008833.2 - 2 254 4304656
HY021361.1 - 2 269

View Full Product Details