Gene Transcripts

Gene Symbol
Npepo
Entrez Gene ID
290963
Gene Name
aminopeptidase O
Gene Aliases
Aopep, Apo, RGD1309592
Chromosome Location
Chr. 17: 507359 - 825062 on Build Rnor_6.0
UniGene ID
Rn.229038
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012346.1 NP_001012346.1 1 282
XM_008771429.1 XP_008769651.1 3 730
XM_008771430.1 XP_008769652.1 3 730
XM_008771431.1 XP_008769653.1 2 577
XM_008771432.1 XP_008769654.1 2 569
XM_008771433.1 XP_008769655.1 2 442
GenBank mRNA AJ810421.1 - 1 282

View Full Product Details