Gene Transcripts

Gene Symbol
Fam26e
Entrez Gene ID
294431
Gene Name
family with sequence similarity 26, member E
Gene Aliases
RGD1310405
Chromosome Location
Chr. 20: 27651817 - 27657895 on Build Rnor_6.0
UniGene ID
Rn.104918
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024977.1 NP_001020148.1 2 777
GenBank mRNA BC089225.1 - 2 777

View Full Product Details