Gene Transcripts

Gene Symbol
OR6C4
Entrez Gene ID
341418
Gene Name
olfactory receptor family 6 subfamily C member 4
Gene Aliases
OR12-10
Chromosome Location
Chr. 12: 55551227 - 55552156 on Build GRCh38
UniGene ID
Hs.495021
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005494.1 NP_001005494.1 1 63
GenBank mRNA BC137000.1 - 1 91
BC137001.1 - 1 91 9020993

View Full Product Details