Gene Transcripts

Gene Symbol
OR4A47
Entrez Gene ID
403253
Gene Name
olfactory receptor family 4 subfamily A member 47
Gene Aliases
OR11-113
Chromosome Location
Chr. 11: 48488793 - 48489722 on Build GRCh38
UniGene ID
Hs.554532
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005512.2 NP_001005512.2 1 649
GenBank mRNA BC140738.1 - 1 725

View Full Product Details