Gene Transcripts

Gene Symbol
OR5K4
Entrez Gene ID
403278
Gene Name
olfactory receptor family 5 subfamily K member 4
Gene Aliases
Chromosome Location
Chr. 3: 98353854 - 98354819 on Build GRCh38
UniGene ID
Hs.554710
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005517.1 NP_001005517.1 1 581

View Full Product Details