Gene Transcripts

Gene Symbol
CISD2
Entrez Gene ID
493856
Gene Name
CDGSH iron sulfur domain 2
Gene Aliases
ERIS, Miner1, NAF-1, WFS2, ZCD2
Chromosome Location
Chr. 4: 102868978 - 102892807 on Build GRCh38
UniGene ID
Hs.444955
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008388.4 NP_001008389.1 3 1382
GenBank mRNA BC032300.1 - 3 1322
BX537971.1 - 3 1320
DB565768.1 - 1 189

View Full Product Details