Gene Transcripts

Gene Symbol
Vom1r62
Entrez Gene ID
494289
Gene Name
vomeronasal 1 receptor 62
Gene Aliases
V1rk2
Chromosome Location
Chr. 1: 75326217 - 75327164 on Build Rnor_6.0
UniGene ID
Rn.129407
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008943.1 NP_001008943.1 1 298
GenBank mRNA AY510309.1 - 1 298

View Full Product Details