Gene Transcripts

Gene Symbol
Spetex-2A
Entrez Gene ID
364261
Gene Name
Spetex-2A protein
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.198259
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009976.1 NP_001009976.1 4 635
GenBank mRNA AB180076.1 - 4 635

View Full Product Details