Gene Transcripts

Gene Symbol
Pld3
Entrez Gene ID
361527
Gene Name
phospholipase D family, member 3
Gene Aliases
Chromosome Location
Chr. 1: 84339269 - 84361686 on Build Rnor_6.0
UniGene ID
Rn.128036
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012167.1 NP_001012167.1 12 1466
XM_006228578.2 XP_006228640.1 14 1795
XM_006228579.2 XP_006228641.1 14 1743
XM_006228580.2 XP_006228642.1 13 1901
XM_006228581.2 XP_006228643.1 14 1773
XM_008759165.1 XP_008757387.1 13 1686
GenBank mRNA BC089987.1 - 12 1466

View Full Product Details