Gene Transcripts

Gene Symbol
TAS2R19
Entrez Gene ID
259294
Gene Name
taste 2 receptor member 19
Gene Aliases
MSTP058, T2R19, T2R23, T2R48, TAS2R23, TAS2R48
Chromosome Location
Chr. 12: 11021619 - 11022620 on Build GRCh38
UniGene ID
Hs.687025
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176888.2 NP_795369.1 1 407
GenBank mRNA BC101804.1 - 1 407
BC111998.1 - 1 419 8327466

View Full Product Details