Gene Transcripts

Gene Symbol
Vasp
Entrez Gene ID
22323
Gene Name
vasodilator-stimulated phosphoprotein
Gene Aliases
Chromosome Location
Chr. 7: 19256929 - 19271854 on Build GRCm38
UniGene ID
Mm.9684
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282021.1 NP_001268950.1 7 1084
NM_001282022.1 NP_001268951.1 7 1057
NM_009499.3 NP_033525.2 7 1087
GenBank mRNA AF084548.1 - 7 1054
AK140329.1 - 7 1051
AK150103.1 - 7 1049
AK155480.1 - 7 1050
AK157021.1 - 7 1048
AK170780.1 - 7 1013
AK171302.1 - 7 1052
AK171715.1 - 7 1051
BC010223.1 - 7 789 3500587
BC015289.1 - 7 1030 4240907

View Full Product Details