Gene Transcripts
- Gene Symbol
- OR1N1
- Entrez Gene ID
- 138883
- Gene Name
- olfactory receptor family 1 subfamily N member 1
- Gene Aliases
- OR1-26, OR1N3, OR9-22
- Chromosome Location
- Chr. 9: 122526358 - 122527293 on Build GRCh38
- UniGene ID
- Hs.553604
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012363.1 |
NP_036495.1
|
1 |
662 |
|
View Full Product Details