Gene Transcripts

Gene Symbol
WDR13
Entrez Gene ID
64743
Gene Name
WD repeat domain 13
Gene Aliases
MG21
Chromosome Location
Chr. X: 48597492 - 48605194 on Build GRCh38
UniGene ID
Hs.521973
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166426.1 NP_001159898.1 8 2177
NM_017883.4 NP_060353.2 9 1842
NR_029427.1 9 1808
XM_006724539.1 XP_006724602.1 10 1495
XM_006724541.3 XP_006724604.1 10 1461
XM_011543947.2 XP_011542249.1 10 1455
GenBank mRNA AF329819.4 - 9 1842
AK000570.1 - 4 488
AK225542.1 - 4 488
AK314739.1 - 9 1805
BC002507.2 - 6 904
BC067094.1 - 9 1489
BC080579.1 - 9 1571 6644248
BX649101.1 - 7 787
DQ907252.1 - 10 1534
DQ907253.1 - 8 1849
DQ907254.1 - 8 1843
DQ907255.1 - 8 1792

View Full Product Details