Gene Transcripts

Gene Symbol
Trim40
Entrez Gene ID
365528
Gene Name
tripartite motif-containing 40
Gene Aliases
Rnf35
Chromosome Location
Chr. 20: 2169674 - 2178477 on Build Rnor_6.0
UniGene ID
Rn.215808
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009175.1 NP_001009175.1 3 485
XM_008772696.1 XP_008770918.1 3 485

View Full Product Details