Gene Transcripts

Gene Symbol
Lrrc39
Entrez Gene ID
109245
Gene Name
leucine rich repeat containing 39
Gene Aliases
2010005E21Rik, 9430028I06Rik
Chromosome Location
Chr. 3: 116562953 - 116583134 on Build GRCm38
UniGene ID
Mm.216027
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027321.3 NP_081597.1 3 372
NM_175413.3 NP_780622.1 3 372
GenBank mRNA AK008119.1 - 3 340
AK035072.1 - 3 374
AK039277.1 - 3 369
AK046671.1 - 3 268
AK052159.1 - 3 324
AK079125.1 - 3 350
BC119529.2 - 3 321 40056120
BC119530.1 - 3 322 40056121

View Full Product Details