Gene Transcripts

Gene Symbol
PTCD3
Entrez Gene ID
55037
Gene Name
pentatricopeptide repeat domain 3
Gene Aliases
MRP-S39
Chromosome Location
Chr. 2: 86106182 - 86142157 on Build GRCh38
UniGene ID
Hs.323489
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017952.5 NP_060422.4 23, 24 2054
GenBank mRNA AK000765.1 - 13, 14 1225
AK315676.1 - 17, 18 1508
AL832520.1 - 12, 13 1058
AY277598.1 - 11, 12 1022
BC001758.1 - 12, 13 1128
BC011832.2 - 23, 24 1989 3939273

View Full Product Details