Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013414.3 | NP_001013432.1 | 15 | 2521 | |
NM_001291118.1 | NP_001278047.1 | 15 | 2521 | ||
NM_001291119.1 | NP_001278048.1 | 14 | 2455 | ||
GenBank mRNA | AK157107.1 | - | 14 | 2321 | |
AK173264.1 | - | 14 | 2016 | ||
BC158107.1 | - | 15 | 2521 | 9007389 | |
BC172172.1 | - | 14 | 2455 | 9054842 |