Gene Transcripts

Gene Symbol
Parp16
Entrez Gene ID
315760
Gene Name
poly (ADP-ribose) polymerase family, member 16
Gene Aliases
ARTD15, LRRGT00109, RGD1306243
Chromosome Location
Chr. 8: 70712392 - 70731212 on Build Rnor_6.0
UniGene ID
Rn.23423
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014093.1 NP_001014115.1 2 380
XM_006243257.2 XP_006243319.1 3 768
XM_006243258.2 XP_006243320.1 3 536
XM_008766279.1 XP_008764501.1 3 768
GenBank mRNA AY387095.1 - 632
BC085908.1 - 2 380 7096186

View Full Product Details