Gene Transcripts

Gene Symbol
VPS13C
Entrez Gene ID
54832
Gene Name
vacuolar protein sorting 13 homolog C
Gene Aliases
PARK23
Chromosome Location
Chr. 15: 61852389 - 62060465 on Build GRCh38
UniGene ID
Hs.511668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018088.2 NP_001018098.1 60, 61 8154
NM_017684.4 NP_060154.3 58, 59 8025
NM_018080.3 NP_060550.2 58, 59 8025
NM_020821.2 NP_065872.1 60, 61 8154
XM_011521713.2 XP_011520015.1 60, 61 8115
GenBank mRNA AJ608770.1 - 58, 59 8008
AJ608771.1 - 60, 61 8137
AJ626860.1 - 58, 59 8008
AJ626861.1 - 60, 61 8137
AK001243.1 - 1, 2 200
AK128650.1 - 14, 15 2568
CR936716.1 - 9, 10 1635

View Full Product Details